2-Piperazin-1-yl-1-piperidin-1-ylethanone 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR0240-5G
OR0240-250MG
OR0240-500MG
OR0240-1G
OR0240-2.5G
APOSOR0240-5GEA
4470
CZK
APOSOR0240-5G
APOSOR0240-250MG
APOSOR0240-500MG
APOSOR0240-1G
APOSOR0240-2.5G
2-Piperazin-1-yl-1-piperidin-1-ylethanone 97%
2-Piperazin-1-yl-1-piperidin-1-ylethanone
Vzorec:
C₁₁H₂₁N₃O MW: 211,31 g/mol Bod varu: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075233 CAS číslo: 70558-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...