Tisk…

(4-Pyridylmethyl)hydrazine dihydrochloride 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(Hydrazinylmethyl)pyridine dihydrochloride, 4-(Hydrazinomethyl)pyridine dihydrochloride

OR0207-1G OR0207-250MG
APOSOR0207-1GEA 1990 CZK
APOSOR0207-1G APOSOR0207-250MG
(4-Pyridylmethyl)hydrazine dihydrochloride 95+%
(4-Pyridylmethyl)hydrazine dihydrochloride
Vzorec: C₆H₁₁Cl₂N₃
MW: 196,08 g/mol
Bod tání: >200 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD06245508
CAS číslo: 89598-56-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR