Ethyl-2-(methylamino)-1,3-thiazole-4-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR01961-1G
OR01961-250MG
OR01961-5G
APOSOR01961-1GEA
3810
CZK
APOSOR01961-1G
APOSOR01961-250MG
APOSOR01961-5G
Ethyl-2-(methylamino)-1,3-thiazole-4-carboxylate
Ethyl-2-(methylamino)-1,3-thiazole-4-carboxylate
Vzorec:
C₇H₁₀N₂O₂S MW: 186,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06251745 CAS číslo: 57250-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...