Tisk…

tert-Butoxy acetic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(tert-Butoxy)acetic acid

OR0183-25G OR0183-5G OR0183-100G
APOSOR0183-25GEA 4240 CZK
APOSOR0183-25G APOSOR0183-5G APOSOR0183-100G
tert-Butoxy acetic acid 98%
tert-Butoxy acetic acid
Vzorec: C₆H₁₂O₃
MW: 132,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03452738
CAS číslo: 13211-32-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR