Tisk…

Ethyl-4-methylbenzoylformate, technická kvalita

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2-(4-methylphenyl)glyoxylate, Ethyl 4-Methylbenzoylformate

OR017808-1G OR017808-5G
APOSOR017808-1GEA 1950 CZK
APOSOR017808-1G APOSOR017808-5G
Ethyl-4-methylbenzoylformate, technická kvalita
Ethyl-4-methylbenzoylformate
Vzorec: C₁₁H₁₂O₃
MW: 192,2100 g/mol
Bod varu: 90…92 °C/0,2mmHg
Storage Temperature: Ambient
MDL Number: MFCD01311169
CAS číslo: 5524-56-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR