Methyl-2-(piperidin-4-ylsulphonyl)benzoate hydrochloride 95+%
Dodavatel: Apollo Scientific
OR0173-1G
OR0173-5G
APOSOR0173-1GEA
8100
CZK
APOSOR0173-1G
APOSOR0173-5G
Methyl-2-(piperidin-4-ylsulphonyl)benzoate hydrochloride 95+%
Methyl-2-(piperidin-4-ylsulphonyl)benzoate hydrochloride
Vzorec:
C₁₃H₁₈ClNO₄S MW: 319,81 g/mol Bod tání: 225…227 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115825 CAS číslo: 849035-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...