Tisk…

a-Tosyl(4-bromobenzyl)isocyanide 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Bromophenyl)(isocyano)methyl 4-methylphenyl sulphone

OR01701-1G OR01701-250MG
APOSOR01701-1GEA 9260 CZK
APOSOR01701-1G APOSOR01701-250MG
a-Tosyl(4-bromobenzyl)isocyanide 97%
a-Tosyl(4-bromobenzyl)isocyanide
Vzorec: C₁₅H₁₂BrNO₂S
Storage Temperature: Ambient
CAS číslo: 655254-61-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR