Tisk…

1-N-Boc-4-(4-pyridylmethylamino)piperidine 95+%

Dodavatel: Apollo Scientific

Synonyms: 4-[(Pyridin-4-yl)methylamino]piperidine, N1-BOC protected

OR01645-1G OR01645-5G OR01645-25G
APOSOR01645-1GEA 6450 CZK
APOSOR01645-1G APOSOR01645-5G APOSOR01645-25G
1-N-Boc-4-(4-pyridylmethylamino)piperidine 95+%
1-N-Boc-4-(4-pyridylmethylamino)piperidine
Vzorec: C₁₆H₂₅N₃O₂
MW: 291,4 g/mol
Bod tání: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD04114985
CAS číslo: 206274-24-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR