Tisk…

(S)-(+)-2-Aminobutyric acid

Dodavatel: Apollo Scientific
Varování

Synonyms: H-Abu-OH, L-alpha-Amino-N-Butyric acid, L-2-Aminobutyric acid, (S)-2-Aminobutanoic acid

OR01636-100G OR01636-25G OR01636-5G
APOSOR01636-100GEA 1040 CZK
APOSOR01636-100G APOSOR01636-25G APOSOR01636-5G
(S)-(+)-2-Aminobutyric acid
(S)-(+)-2-Aminobutyric acid
Vzorec: C₄H₉NO₂
MW: 103,12 g/mol
Bod tání: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064415
CAS číslo: 1492-24-6
EINECS: 216-083-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR