Tisk…

Terephthalamic acid 95+%

Dodavatel: Apollo Scientific
Varování

OR016303-1G OR016303-5G
APOSOR016303-1GEA 705 CZK
APOSOR016303-1G APOSOR016303-5G
Terephthalamic acid 95+%
Terephthalamic acid
Vzorec: C₈H₇NO₃
Bod tání: >330 °C
Storage Temperature: Ambient
MDL Number: MFCD00025482
CAS číslo: 6051-43-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR