3-Chloroadamantane-1-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR01607-1G
OR01607-5G
OR01607-10G
APOSOR01607-1GEA
1580
CZK
APOSOR01607-1G
APOSOR01607-5G
APOSOR01607-10G
3-Chloroadamantane-1-carboxylic acid
3-Chloroadamantane-1-carboxylic acid
Vzorec:
C₁₁H₁₅ClO₂ Bod tání: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167831 CAS číslo: 34859-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...