1-Phenyl-2-(pyrimidin-4-yl)ethanol 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR0157-1G
OR0157-5G
APOSOR0157-1GEA
2070
CZK
APOSOR0157-1G
APOSOR0157-5G
1-Phenyl-2-(pyrimidin-4-yl)ethanol 95+%
1-Phenyl-2-(pyrimidin-4-yl)ethanol
Vzorec:
C₁₂H₁₂N₂O MW: 200,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03426053 CAS číslo: 36914-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...