3-Bromoimidazo[1,2-a]pyrazine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR01540-5G
APOSOR01540-5GEA
3230
CZK
APOSOR01540-5G
3-Bromoimidazo[1,2-a]pyrazine
3-Bromoimidazo[1,2-a]pyrazine
Vzorec:
C6H4BrN3 MW: 198,0200 g/mol Bod tání: 191…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD06245372 CAS číslo: 57948-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...