Tisk…

1-(4-Methoxyphenyl)-2-(quinoxalin-2-yl)ethanol 95+%

Dodavatel: Apollo Scientific

OR0154-10G OR0154-1G OR0154-5G
APOSOR0154-10GEA 17260 CZK
APOSOR0154-10G APOSOR0154-1G APOSOR0154-5G
1-(4-Methoxyphenyl)-2-(quinoxalin-2-yl)ethanol 95+%
1-(4-Methoxyphenyl)-2-(quinoxalin-2-yl)ethanol
Vzorec: C₁₇H₁₆N₂O₂
MW: 280.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426056
CAS číslo: 849021-36-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR