Tisk…

1-(4-Methoxyphenyl)-2-(quinolin-2-yl)ethanol 95+%

Dodavatel: Apollo Scientific

OR0153-1G OR0153-5G
APOSOR0153-1GEA 4630 CZK
APOSOR0153-1G APOSOR0153-5G
1-(4-Methoxyphenyl)-2-(quinolin-2-yl)ethanol 95+%
1-(4-Methoxyphenyl)-2-(quinolin-2-yl)ethanol
Vzorec: C₁₈H₁₇NO₂
MW: 279,34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03452779
CAS číslo: 94004-77-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR