3-Ethynylbenzaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR01515-1G
OR01515-5G
OR01515-10G
APOSOR01515-1GEA
2280
CZK
APOSOR01515-1G
APOSOR01515-5G
APOSOR01515-10G
3-Ethynylbenzaldehyde 97%
3-Ethynylbenzaldehyde
Vzorec:
C₉H₆O MW: 130.1433 g/mol Bod tání: 75…80 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06245349 CAS číslo: 77123-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...