2-Ethynylaniline 95%
Dodavatel: Apollo Scientific
Synonyms:
2-Aminophenylacetylene
OR015038-10G
OR015038-1G
OR015038-5G
APOSOR015038-10GEA
12700
CZK
APOSOR015038-10G
APOSOR015038-1G
APOSOR015038-5G
2-Ethynylaniline 95%
2-Ethynylaniline
Vzorec:
C₈H₇N Bod varu: 229…230 °C Hustota: 1,03 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00168862 CAS číslo: 52670-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...