3,4-Dichlorophenyl isocyanate 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR015005-100G
OR015005-25G
OR015005-500G
APOSOR015005-100GEA
2690
CZK
APOSOR015005-100G
APOSOR015005-25G
APOSOR015005-500G
3,4-Dichlorophenyl isocyanate 97%
3,4-Dichlorophenyl isocyanate
When melted, may contain small amount of sediment.
Vzorec:
C₇H₃Cl₂NO MW: 188,01 g/mol Bod varu: 118…120 °C Bod tání: 40…44 °C Bod vzplanutí: 123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002017 CAS číslo: 102-36-3 UN: 2250 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...