Tisk…

3-Phenylpyrazole-5-carboxylic acid hydrate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Phenyl-1H-pyrazole-3-carboxylic acid

OR01491-1G OR01491-250MG OR01491-5G
APOSOR01491-1GEA 1240 CZK
APOSOR01491-1G APOSOR01491-250MG APOSOR01491-5G
3-Phenylpyrazole-5-carboxylic acid hydrate 97%
3-Phenylpyrazole-5-carboxylic acid hydrate
Vzorec: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Bod tání: 250…258 °C
Storage Temperature: Ambient
MDL Number: MFCD01248821
CAS číslo: 5071-61-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR