3-(1-Adamantyl)propanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(Adamantan-1-yl)propanoic acid
OR01478-1G
OR01478-250MG
APOSOR01478-1GEA
9100
CZK
APOSOR01478-1G
APOSOR01478-250MG
3-(1-Adamantyl)propanoic acid
3-(1-Adamantyl)propanoic acid
Vzorec:
C₁₃H₂₀O₂ MW: 208,3 g/mol Bod tání: 147…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00194029 CAS číslo: 16269-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...