Tisk…

5-Chloro-2-iodotoluene

Dodavatel: Apollo Scientific
Varování

OR0133-100G
APOSOR0133-100GEA 2570 CZK
APOSOR0133-100G
5-Chloro-2-iodotoluene
5-Chloro-2-iodotoluene
Vzorec: C₇H₆ClI
MW: 252,48 g/mol
Bod varu: 120…122 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00060665
CAS číslo: 23399-70-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR