Tisk…

1-(2-Pyridin-2-ylethyl)piperazine 97%

Dodavatel: Apollo Scientific
Varování

OR0068-1G OR0068-5G
APOSOR0068-1GEA 2690 CZK
APOSOR0068-1G APOSOR0068-5G
1-(2-Pyridin-2-ylethyl)piperazine 97%
1-(2-Pyridin-2-ylethyl)piperazine
Vzorec: C₁₁H₁₇N₃
Storage Temperature: Ambient
MDL Number: MFCD01748645
CAS číslo: 53345-15-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR