(2,4-Dimethoxyphenyl)acetic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,4-Dimethoxyphenylacetic Acid
OR0056-100G
OR0056-25G
OR0056-5G
APOSOR0056-100GEA
9560
CZK
APOSOR0056-100G
APOSOR0056-25G
APOSOR0056-5G
(2,4-Dimethoxyphenyl)acetic acid 98%
(2,4-Dimethoxyphenyl)acetic acid
Vzorec:
(C₆H₃(OCH₃)₂)CH₂COOH MW: 196,2 g/mol Bod tání: 108…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00183328 CAS číslo: 6496-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...