Tisk…

2,3,4-Trichlorobenzenesulphonyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,3,4-Trichlorobenzene-1-sulphonyl chloride

OR0054-100G OR0054-25G OR0054-5G
APOSOR0054-100GEA 53230 CZK
APOSOR0054-100G APOSOR0054-25G APOSOR0054-5G
2,3,4-Trichlorobenzenesulphonyl chloride 98%
2,3,4-Trichlorobenzenesulphonyl chloride
Vzorec: C₆H₂Cl₄O₂S
MW: 279,96 g/mol
Bod tání: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00024871
CAS číslo: 34732-09-7
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR