3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,5-Di-tert-butylsalicylaldehyde
OR0037-100G
OR0037-25G
OR0037-500G
APOSOR0037-100GEA
705
CZK
APOSOR0037-100G
APOSOR0037-25G
APOSOR0037-500G
3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%
3,5-Di-tert-butyl-2-hydroxybenzaldehyde
Vzorec:
C₁₅H₂₂O₂ MW: 234.34 g/mol Bod tání: 59…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191998 CAS číslo: 37942-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...