Tisk…

Zirconium(IV) isopropoxide isopropanol complex

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Zirconium(IV) i-propoxide (isopropanol adduct)

IN3919-10G IN3919-100G
APOSIN3919-10GEA 4470 CZK
APOSIN3919-10G APOSIN3919-100G
Zirconium(IV) isopropoxide isopropanol complex
Zirconium(IV) isopropoxide isopropanol complex
Vzorec: C₁₅H₃₆O₅Zr
MW: 387.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075157
CAS číslo: 14717-56-7
UN: 3181
ADR: 4.1 II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR