Dusičnan-oxid zirkoničitý hydrát
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate
IN3913-250G
IN3913-50G
APOSIN3913-250GEA
18190
CZK
APOSIN3913-250G
APOSIN3913-50G
Dusičnan-oxid zirkoničitý hydrát
Dusičnan-oxid zirkoničitý hydrát
Vzorec:
N₂O₇Zr·H₂O MW: 249.25 Storage Temperature: Ambient |
MDL Number:
MFCD00149896 CAS číslo: 14985-18-3 EINECS: 237-529-3 UN: 3085 ADR: 5.1+8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...