Tisk…

Vanadium(III) chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

IN3763-100G IN3763-25G IN3763-5G
APOSIN3763-100GEA 11410 CZK
APOSIN3763-100G APOSIN3763-25G APOSIN3763-5G
Vanadium(III) chloride 97%
Vanadium(III) chloride
Vzorec: Cl₃V
Bod tání: 250 °C
Hustota: 3 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011454
CAS číslo: 7718-98-1
UN: 2475
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR