Tisk…

Terbium(III) chloride, anhydrous 99,9%

Dodavatel: Apollo Scientific
Varování

IN3475-1G IN3475-5G
APOSIN3475-1GEA 3110 CZK
APOSIN3475-1G APOSIN3475-5G
Terbium(III) chloride, anhydrous 99,9%
Terbium(III) chloride
Vzorec: Cl₃Tb
MW: 265.28
Bod tání: 588 °C
Hustota: 4.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011258
CAS číslo: 10042-88-3
EINECS: 233-132-4
Merck Index: 13,09087

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR