Tisk…

Tantalum(V) ethanolate 99.99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Tantalum(V) ethoxide

IN3409-10G IN3409-50G
APOSIN3409-10GEA 4570 CZK
APOSIN3409-10G APOSIN3409-50G
Tantalum(V) ethanolate 99.99%
Tantalum(V) ethanolate
Vzorec: C₁₀H₂₅O₅Ta
Bod tání: 21 °C
Hustota: 1,566 g/cm³ (25 °C)
Bod vzplanutí: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00049785
CAS číslo: 6074-84-6
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR