Strontium oxide 99,5%
Dodavatel: Apollo Scientific
IN3373-100G
IN3373-25G
APOSIN3373-100GEA
15740
CZK
APOSIN3373-100G
APOSIN3373-25G
Strontium oxide 99,5%
Strontium oxide
Vzorec:
SrO Bod tání: 2430 °C Hustota: 4,7 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049558 CAS číslo: 1314-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...