Tisk…

Sodium thiophosphate hydrate

Dodavatel: Apollo Scientific
Varování

IN3322-25G IN3322-1G IN3322-5G
APOSIN3322-25GEA 7250 CZK
APOSIN3322-25G APOSIN3322-1G APOSIN3322-5G
Sodium thiophosphate hydrate
Sodium thiophosphate hydrate
Vzorec: H₂Na₃O₄PS
Bod tání: 56…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041513
CAS číslo: 10489-48-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR