Tisk…

Oxid skanditý 99,9%

Dodavatel: Apollo Scientific

Synonyms: Scandium oxide, Scandium (III) oxide

IN3145-1G IN3145-5G
APOSIN3145-1GEA 2200 CZK
APOSIN3145-1G APOSIN3145-5G
Oxid skanditý 99,9%
Oxid skanditý
Vzorec: O₃Sc₂
MW: 137,91 g/mol
Hustota: 3,864 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011223
CAS číslo: 12060-08-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR