Tisk…

Scandium(III) chloride, anhydrous 99,9%

Dodavatel: Apollo Scientific
Varování

IN3133-1G IN3133-5G
APOSIN3133-1GEA 8900 CZK
APOSIN3133-1G APOSIN3133-5G
Scandium(III) chloride, anhydrous 99,9%
Scandium(III) chloride
Vzorec: Cl₃Sc
MW: 151.31
Bod tání: 960 °C
Hustota: 2.39 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011221
CAS číslo: 10361-84-9
EINECS: 233-799-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR