Praseodymium(III,IV) oxide 99,9%
Dodavatel: Apollo Scientific
IN2980-250G
IN2980-50G
APOSIN2980-250GEA
7670
CZK
APOSIN2980-250G
APOSIN2980-50G
Praseodymium(III,IV) oxide 99,9%
Praseodymium(III,IV) oxide
Vzorec:
O₁₁Pr₆ MW: 1021,44 g/mol Bod varu: 4200 °C Bod tání: 2500 °C Hustota: 7,07 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011178 CAS číslo: 12037-29-5 EINECS: 234-857-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...