Tisk…

Praseodymium trifluoride 99,9%

Dodavatel: Apollo Scientific

IN2971-100G IN2971-25G
APOSIN2971-100GEA 30780 CZK
APOSIN2971-100G APOSIN2971-25G
Praseodymium trifluoride 99,9%
Praseodymium trifluoride
Vzorec: F₃Pr
MW: 197,9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016287
CAS číslo: 13709-46-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR