Tisk…

Niobium(V) chloride 99,9%

Dodavatel: Apollo Scientific
Nebezpečí

IN2743-100G IN2743-25G
APOSIN2743-100GEA 13660 CZK
APOSIN2743-100G APOSIN2743-25G
Niobium(V) chloride 99,9%
Niobium(V) chloride
Vzorec: Cl₅Nb
Bod varu: 254 °C
Hustota: 2,75 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011127
CAS číslo: 10026-12-7
UN: 3260
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR