Chlorid nikelnatý, anhydrous 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Nickel dichloride
IN2677-250G
IN2677-50G
APOSIN2677-250GEA
9920
CZK
APOSIN2677-250G
APOSIN2677-50G
Chlorid nikelnatý, anhydrous 98%
Chlorid nikelnatý
Vzorec:
Cl₂Ni MW: 129,6 g/mol Bod tání: 1001 °C Hustota: 3,55 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011142 CAS číslo: 7718-54-9 EINECS: 231-743-0 UN: 3288 ADR: 6.1,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...