Oxid neodymitý 99,9%
Dodavatel: Apollo Scientific
Synonyms:
Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
IN2650-250G
IN2650-50G
APOSIN2650-250GEA
5790
CZK
APOSIN2650-250G
APOSIN2650-50G
Oxid neodymitý 99,9%
Oxid neodymitý
Vzorec:
Nd₂O₃ MW: 336.48 Bod varu: 3760 °C Bod tání: 2272 °C Hustota: 7.24 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011134 CAS číslo: 1313-97-9 EINECS: 215-214-1 Merck Index: 14,06450 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...