Holmium 99,9%, powder –40 mesh
Dodavatel: Apollo Scientific
IN1966-25G
IN1966-5G
APOSIN1966-25GEA
41330
CZK
APOSIN1966-25G
APOSIN1966-5G
Holmium 99,9%, powder –40 mesh
Holmium
Vzorec:
Ho Bod varu: 2600 °C Bod tání: 1474 °C Hustota: 8,8 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011049 CAS číslo: 7440-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...