Tisk…

Gallium tribromide, anhydrous 99.99%

Dodavatel: Apollo Scientific

Synonyms: Gallium (III) bromide

IN1867-100G IN1867-25G
APOSIN1867-100GEA 43640 CZK
APOSIN1867-100G APOSIN1867-25G
Gallium tribromide, anhydrous 99.99%
Gallium tribromide
Vzorec: Br₃Ga
Bod varu: 279 °C
Storage Temperature: Ambient
MDL Number: MFCD00016100
CAS číslo: 13450-88-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR