Europium(III) bromide 99.99%
Dodavatel: Apollo Scientific
IN1762-100G
IN1762-10G
APOSIN1762-100GEA
29760
CZK
APOSIN1762-100G
APOSIN1762-10G
Europium(III) bromide 99.99%
Europium(III) bromide
Vzorec:
Br₃Eu Bod tání: 702 °C Storage Temperature: Ambient |
CAS číslo:
13759-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...