Tisk…

Erbium trichloride hexahydrate 99.99%

Dodavatel: Apollo Scientific
Varování

IN1696-100G IN1696-25G
APOSIN1696-100GEA 7670 CZK
APOSIN1696-100G APOSIN1696-25G
Erbium trichloride hexahydrate 99.99%
Erbium trichloride hexahydrate
Vzorec: Cl₃Er·6H₂O
MW: 381.71
Bod tání: 774 °C
Storage Temperature: Ambient
MDL Number: MFCD00149693
CAS číslo: 10025-75-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR