Erbium trichloride, anhydrous 99,9%
Dodavatel: Apollo Scientific
|
Varování
|
IN1695-100G
IN1695-25G
APOSIN1695-100GEA
10910
CZK
APOSIN1695-100G
APOSIN1695-25G
Erbium trichloride, anhydrous 99,9%
Erbium trichloride
Vzorec:
Cl₃Er Bod varu: 1500 °C Bod tání: 774 °C Hustota: 4,1 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010989 CAS číslo: 10138-41-7 EINECS: 233-385-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...