Dysprosium trinitrate pentahydrate 99,9%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Dysprosium (III) nitrate pentahydrate
IN1651-100G
IN1651-25G
APOSIN1651-100GEA
14220
CZK
APOSIN1651-100G
APOSIN1651-25G
Dysprosium trinitrate pentahydrate 99,9%
Dysprosium trinitrate pentahydrate
Vzorec:
DyN₃O₉·5H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149683 CAS číslo: 10031-49-9 EINECS: 233-410-5 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...