Copper selenide 99.99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Copper(II) selenide
IN1585-100G
IN1585-25G
APOSIN1585-100GEA
17590
CZK
APOSIN1585-100G
APOSIN1585-25G
Copper selenide 99.99%
Copper selenide
Vzorec:
CuSe MW: 142,51 g/mol Bod tání: 550 °C (rozkl.) Hustota: 5,99 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016058 CAS číslo: 1317-41-5 EINECS: 215-272-8 UN: 3283 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...