Tisk…
Copper(II) acetate 96%
Dodavatel: Apollo Scientific
Nebezpečí
Synonyms: Acetic acid copper (II) salt, Cupric acetate, Copper diacetate
IN1535-25G IN1535-50G
APOSIN1535-25GEA 622 CZK
APOSIN1535-25G APOSIN1535-50G
Copper(II) acetate 96%
Copper(II) acetate
Vzorec: CuCH₃CO₂
MW: 181.64
Bod varu: 240
Bod tání: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00008690
CAS číslo: 142-71-2
EINECS: 205-553-3
UN: 3077
ADR: 9+EHS III
Merck Index: 12,02690