Tisk…

Chromium(III) nitrate 99,9%

Dodavatel: Apollo Scientific
Varování

IN1477-100G IN1477-25G
APOSIN1477-100GEA 13460 CZK
APOSIN1477-100G APOSIN1477-25G
Chromium(III) nitrate 99,9%
Chromium(III) nitrate
Vzorec: CrN₃O₉
MW: 238,01 g/mol
Bod tání: 60 °C
Storage Temperature: Ambient
CAS číslo: 13548-38-4
UN: 2720
ADR: 5.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR