Tisk…

Antimony (III) ethanolate 99.99+%

Dodavatel: Apollo Scientific

IN1117-50G IN1117-10G
APOSIN1117-50GEA 26060 CZK
APOSIN1117-50G APOSIN1117-10G
Antimony (III) ethanolate 99.99+%
Antimony (III) ethanolate
Vzorec: C₆H₁₅O₃Sb
Bod varu: 95 °C/11mmHg
Hustota: 1,524 g/cm³ (25 °C)
Bod vzplanutí: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00015123
CAS číslo: 10433-06-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR