Ammonium tetrathiomolybdate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ammonium tetrathiomolybdate(VI)
IN1087-10G
IN1087-50G
IN1087-1G
APOSIN1087-10GEA
8700
CZK
APOSIN1087-10G
APOSIN1087-50G
APOSIN1087-1G
Ammonium tetrathiomolybdate
Ammonium tetrathiomolybdate
Vzorec:
H₁₀MoN₂S₄ MW: 262.284 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00136013 CAS číslo: 15060-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...